accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3CH3OCH3CH3CH3OZn

CAS RN 4278-43-7

IUPAC Name: zinc(2+) bis(2-methylpropan-2-olate)
Synonyms: zinc(2+) bis(2-methylpropan-2-olate)
Molecular Weight (g/mol): 211.61
Molecular Formula: C8H18O2Zn
InChi Key: DVGVEVPHJQJMPP-UHFFFAOYSA-N
SMILES: [Zn++].CC(C)(C)[O-].CC(C)(C)[O-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results