accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 427896-41-1

CH3SNH3CCH3H3COOH

CAS RN 427896-41-1

IUPAC Name: 2-tert-butyl-4-methyl-1,3-thiazole-5-carboxylic acid
Synonyms: 2-tert-butyl-4-methyl-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 199.27
Molecular Formula: C9H13NO2S
InChi Key: YHLQWXNPKLIQMJ-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results