accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4282-31-9

IUPAC Name: thiophene-2,5-dicarboxylic acid
Synonyms: 2,5-thiophenedicarboxylic acid
Molecular Weight (g/mol): 172.15
Molecular Formula: C6H4O4S
InChi Key: YCGAZNXXGKTASZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results