accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4287-20-1

H3N(R)OHO

CAS RN 4287-20-1

IUPAC Name: (2R)-2-hydroxy-3-phenoxypropan-1-aminium
Synonyms: (2R)-2-hydroxy-3-phenoxypropan-1-aminium
Molecular Weight (g/mol): 168.22
Molecular Formula: C9H14NO2
InChi Key: JZEHWMUIAKALDN-MRVPVSSYSA-O
SMILES: [NH3+]C[C@@H](O)COC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results