accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 42899-76-3

IUPAC Name: pyridine-3-sulfonyl chloride hydrochloride
Synonyms: pyridine-3-sulfonyl chloride hydrochloride
Molecular Weight (g/mol): 214.06
Molecular Formula: C5H5Cl2NO2S
InChi Key: VIVPWOMJFLICOZ-UHFFFAOYSA-N
SMILES: Cl.ClS(=O)(=O)C1=CN=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)