accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FH3CNCH3H3CH3C

CAS RN 429-41-4

IUPAC Name: tetrabutylazanium fluoride
Synonyms: tetra-n-butylammonium fluoride
Molecular Weight (g/mol): 261.47
Molecular Formula: C16H36FN
InChi Key: FPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES: [F-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (15)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results