accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 42918-86-5

CH3HNOOOHO

CAS RN 42918-86-5

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}butanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: SZQMTCSQWUYUML-UHFFFAOYNA-N
SMILES: CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)