accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 42956-75-2

IUPAC Name: N'-hydroxy-2,2-dimethylpropanimidamide
Synonyms: N'-hydroxy-2,2-dimethylpropanimidamide
Molecular Weight (g/mol): 116.16
Molecular Formula: C5H12N2O
InChi Key: CVOGFMYWFRFWEQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results