accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(R)OHHC

CAS RN 42969-65-3

IUPAC Name: (2R)-but-3-yn-2-ol
Synonyms: (2R)-but-3-yn-2-ol
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: GKPOMITUDGXOSB-SCSAIBSYSA-N
SMILES: C[C@@H](O)C#C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)