CAS RN 43038-45-5
IUPAC Name:
naphthalene-1-carbohydrazide
Synonyms:
naphthalene-1-carbohydrazide
Molecular Weight (g/mol):
186.21
Molecular Formula:
C11H10N2O
InChi Key:
VMFUMDXVTKTZQY-UHFFFAOYSA-N
SMILES:
NNC(=O)C1=C2C=CC=CC2=CC=C1
1
–
2
of
2
results
1
1-Naphthoic hydrazide, 98+%
CAS: 43038-45-5 Molecular Formula: C11H10N2O Molecular Weight (g/mol): 186.214 MDL Number: MFCD00016804 InChI Key: VMFUMDXVTKTZQY-UHFFFAOYSA-N Synonym: 1-naphthohydrazide,1-naphthhydrazide,1-naphthalenecarbohydrazide,1-naphthalenecarboxylic acid, hydrazide,1-naphthoic hydrazide,.alpha.-naphthoylhydrazine,naphthalene-1-carboxylic acid hydrazide,naphthalenecarbohydrazide,a-naphthoylhydrazine,1-naphthohydrazide # PubChem CID: 72689 IUPAC Name: naphthalene-1-carbohydrazide SMILES: C1=CC=C2C(=C1)C=CC=C2C(=O)NN