accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiH3CCH3NOO

CAS RN 43112-38-5

IUPAC Name: 3-(trimethylsilyl)-1,3-oxazolidin-2-one
Synonyms: 3-(trimethylsilyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 159.26
Molecular Formula: C6H13NO2Si
InChi Key: AUKCYOUETBBMFV-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N1CCOC1=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)