accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3CNHSOOO

CAS RN 4316-74-9

IUPAC Name: sodium 2-(methylamino)ethane-1-sulfonate
Synonyms: sodium methyltaurate sodium methyl cocoyl taurate
Molecular Weight (g/mol): 161.15
Molecular Formula: C3H8NNaO3S
InChi Key: KKDONKAYVYTWGY-UHFFFAOYSA-M
SMILES: [Na+].CNCCS([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results