accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3NNH

CAS RN 4318-42-7

IUPAC Name: 1-(propan-2-yl)piperazine
Synonyms: piperazine, 1-isopropyl-
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: WHKWMTXTYKVFLK-UHFFFAOYSA-N
SMILES: CC(C)N1CCNCC1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results