accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4319-49-7

IUPAC Name: morpholin-4-amine
Synonyms: morpholin-4-ylamine
Molecular Weight (g/mol): 102.14
Molecular Formula: C4H10N2O
InChi Key: MKQLBNJQQZRQJU-UHFFFAOYSA-N
SMILES: NN1CCOCC1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)