accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4325-85-3

H3CSiH3CCH3OBOSiCH3H3CCH3OSiCH3H3CCH3

CAS RN 4325-85-3

IUPAC Name: tritrimethylsilyl borate
Synonyms: tritrimethylsilyl borate
Molecular Weight (g/mol): 278.38
Molecular Formula: C9H27BO3Si3
InChi Key: YZYKZHPNRDIPFA-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OB(O[Si](C)(C)C)O[Si](C)(C)C
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)