accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4338-95-8

IUPAC Name: hydrogen (methylsulfanyl)methanimidamide iodide
Synonyms: hydrogen methylsulfanylmethanimidamide iodide
Molecular Weight (g/mol): 218.06
Molecular Formula: C2H7IN2S
InChi Key: LFXAECSQJSRSTP-UHFFFAOYSA-N
SMILES: [H+].[I-].CSC(N)=N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results