accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH3CNSNH2N(E)

CAS RN 4338-98-1

IUPAC Name: (2E)-2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloridyl
Synonyms: (2E)-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloridyl
Molecular Weight (g/mol): 214.69
Molecular Formula: C8H9ClN3S
InChi Key: WAGARHVUZQWORV-VRTOBVRTSA-N
SMILES: [Cl].CN1\C(SC2=CC=CC=C12)=N/N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results