accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 434-76-4

IUPAC Name: 2-amino-6-fluorobenzoate
Synonyms: 2-amino-6-fluorobenzoate
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H5FNO2
InChi Key: RWSFZKWMVWPDGZ-UHFFFAOYSA-M
SMILES: NC1=CC=CC(F)=C1C([O-])=O
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results