accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 436151-85-8

IUPAC Name: 4-(2-amino-1,3-thiazol-4-yl)benzonitrile
Synonyms: 4-(2-amino-1,3-thiazol-4-yl)benzonitrile
Molecular Weight (g/mol): 201.25
Molecular Formula: C10H7N3S
InChi Key: KKMZKOIZTSRIEM-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results