accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 436799-33-6

NBrFFF

CAS RN 436799-33-6

IUPAC Name: 3-bromo-5-(trifluoromethyl)pyridine
Synonyms: 3-bromo-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 226
Molecular Formula: C6H3BrF3N
InChi Key: HEDHNDVPKRVQPN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=CN=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)