accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrH3CNCH3CH3CH3

CAS RN 4368-51-8

IUPAC Name: tetraheptylazanium bromide
Synonyms: tetraheptylazanium bromide
Molecular Weight (g/mol): 490.7
Molecular Formula: C28H60BrN
InChi Key: YQIVQBMEBZGFBY-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)