accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ON

CAS RN 4369-55-5

IUPAC Name: 3-phenyl-1,2-oxazol-5-amine
Synonyms: 3-phenyl-1,2-oxazol-5-amine
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: HLOHVVZZMMMDMM-UHFFFAOYSA-N
SMILES: NC1=CC(=NO1)C1=CC=CC=C1
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)