accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 437-64-9

IUPAC Name: 5-hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-chromen-4-one
Synonyms: genkwanin gengkwanin apigenin 7-methyl ether
Molecular Weight (g/mol): 284.27
Molecular Formula: C16H12O5
InChi Key: JPMYFOBNRRGFNO-UHFFFAOYSA-N
SMILES: COC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)