accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 437-81-0

IUPAC Name: 2,6-difluorobenzaldehyde
Synonyms: 2,6-difluorobenzaldehyde
Molecular Weight (g/mol): 142.11
Molecular Formula: C7H4F2O
InChi Key: SOWRUJSGHKNOKN-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1C=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results