accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H2COH

CAS RN 4383-08-8

IUPAC Name: 2-methyl-1-phenylprop-2-en-1-ol
Synonyms: 2-methyl-1-phenylprop-2-en-1-ol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: ZGYBYYJGIKPBFD-UHFFFAOYNA-N
SMILES: CC(=C)C(O)C1=CC=CC=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results