CAS RN 4383-22-6
IUPAC Name:
benzyl(prop-2-en-1-yl)amine
Synonyms:
benzyl(prop-2-en-1-yl)amine
Molecular Weight (g/mol):
147.22
Molecular Formula:
C10H13N
InChi Key:
RHUCQDQRNUUMKY-UHFFFAOYSA-N
SMILES:
C=CCNCC1=CC=CC=C1