accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4385-35-7

IUPAC Name: 3,4-dihydro-1H-2-benzopyran-3-one
Synonyms: 3-isochromanone
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: ILHLUZUMRJQEAH-UHFFFAOYSA-N
SMILES: O=C1CC2=CC=CC=C2CO1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results