accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 438577-61-8

CH3SNFFFO

CAS RN 438577-61-8

IUPAC Name: 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbaldehyde
Synonyms: 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 271.26
Molecular Formula: C12H8F3NOS
InChi Key: TULJSSOBHAIERD-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=C(C=C1)C(F)(F)F)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results