CAS RN 439239-90-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lasmiditan, TRC
CAS: 439239-90-4 Molecular Formula: C19H18F3N3O2 Molecular Weight (g/mol): 377.36 Synonym: 2,4,6-Trifluoro-N-[6-(1-methylpiperidine-4-carbonyl)pyridin-2-yl]benzamide,2,4,6-Trifluoro-N-[6-[(1-methyl-4-piperidinyl)carbonyl]-2-pyridinyl]benzamide SMILES: CN1CCC(CC1)C(=O)c2cccc(NC(=O)c3c(F)cc(F)cc3F)n2