accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCS

CAS RN 4404-45-9

IUPAC Name: 1-isothiocyanatohexane
Synonyms: 1-hexylisothiocyanate
Molecular Weight (g/mol): 143.25
Molecular Formula: C7H13NS
InChi Key: WXYAXKKXIGHXDS-UHFFFAOYSA-N
SMILES: CCCCCCN=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results