accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HONOH(E)

CAS RN 441-38-3

IUPAC Name: (2E)-2-(N-hydroxyimino)-1,2-diphenylethan-1-ol
Synonyms: benzoin oxime
Molecular Weight (g/mol): 227.26
Molecular Formula: C14H13NO2
InChi Key: WAKHLWOJMHVUJC-FYWRMAATNA-N
SMILES: O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)