accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OOH

CAS RN 4410-31-5

IUPAC Name: 2-hydroxy-2-phenylacetamide
Synonyms: O-hydroxy phenylacetamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: MAGPZHKLEZXLNU-UHFFFAOYNA-N
SMILES: NC(=O)C(O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results