accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4411-25-0

IUPAC Name: 1-isocyanatoadamantane
Synonyms: 1-isocyanatoadamantane
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: VBHCPGFCIQDXGZ-UHFFFAOYSA-N
SMILES: O=C=NC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)