accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NHNN

CAS RN 4414-88-4

IUPAC Name: 2-(1H-1,3-benzodiazol-2-yl)acetonitrile
Synonyms: 2-(1H-1,3-benzodiazol-2-yl)acetonitrile
Molecular Weight (g/mol): 157.18
Molecular Formula: C9H7N3
InChi Key: BWOVACANEIVHST-UHFFFAOYSA-N
SMILES: N#CCC1=NC2=CC=CC=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results