accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSiHSOH3COH3C

CAS RN 4420-74-0

IUPAC Name: 3-(trimethoxysilyl)propane-1-thiol
Synonyms: mercaptopropyltrimethoxysilane
Molecular Weight (g/mol): 196.34
Molecular Formula: C6H16O3SSi
InChi Key: UUEWCQRISZBELL-UHFFFAOYSA-N
SMILES: CO[Si](CCCS)(OC)OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results