accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOO

CAS RN 4421-09-4

IUPAC Name: 2H-1,3-benzodioxole-5-carbonitrile
Synonyms: 2H-1,3-benzodioxole-5-carbonitrile
Molecular Weight (g/mol): 147.13
Molecular Formula: C8H5NO2
InChi Key: PKRWWZCDLJSJIF-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2OCOC2=C1
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results