CAS RN 4423-94-3
CAS RN 4423-94-3
IUPAC Name:
2-ethylcyclohexan-1-one
Synonyms:
2-ethylcyclohexan-1-one
Molecular Weight (g/mol):
126.2
Molecular Formula:
C8H14O
InChi Key:
WKYYYUWKFPFVEY-UHFFFAOYNA-N
SMILES:
CCC1CCCCC1=O
1
–
1
of
1
results
1
2-Ethylcyclohexanone, 99%
CAS: 4423-94-3 Molecular Formula: C8H14O Molecular Weight (g/mol): 126.199 MDL Number: MFCD00066306 InChI Key: WKYYYUWKFPFVEY-UHFFFAOYSA-N Synonym: 2-ethylcyclohexanone,cyclohexanone, 2-ethyl,2-ethyl-cyclohexanone,acmc-1advf,2-ethyl-cyclohexan-1-one PubChem CID: 20474 IUPAC Name: 2-ethylcyclohexan-1-one SMILES: CCC1CCCCC1=O