CAS RN 4424-17-3
IUPAC Name:
2-amino-N-phenylbenzamide
Synonyms:
benzamide, 2-amino-N-phenyl-
Molecular Weight (g/mol):
212.25
Molecular Formula:
C13H12N2O
InChi Key:
FDPVTENMNDHFNK-UHFFFAOYSA-N
SMILES:
NC1=CC=CC=C1C(=O)NC1=CC=CC=C1
1
–
1
of
1
results
1
Alfa Aesar™ 2-Aminobenzanilide, 95%
CAS: 4424-17-3 Molecular Formula: C13H12N2O Molecular Weight (g/mol): 212.25 MDL Number: MFCD00034792,MFCD00017096 InChI Key: FDPVTENMNDHFNK-UHFFFAOYSA-N Synonym: 2-aminobenzanilide,benzamide, 2-amino-n-phenyl,anthranilanilide,n-2-aminobenzoyl aniline,n-phenylanthranilamide,benzanilide, 2-amino,n-phenyl-2-aminobenzamide,2-amino-n-phenyl-benzamide,2-aminophenyl-n-benzamide,o-aminobenzanilide PubChem CID: 78142 IUPAC Name: 2-amino-N-phenylbenzamide SMILES: NC1=CC=CC=C1C(=O)NC1=CC=CC=C1