CAS RN 443-48-1
Thermo Scientific Chemicals Metronidazole, 99%
CAS: 443-48-1 Molecular Formula: C6H9N3O3 Molecular Weight (g/mol): 171.16 MDL Number: MFCD00009750 InChI Key: VAOCPAMSLUNLGC-UHFFFAOYSA-N Synonym: metronidazole,metronidazol,flagyl,2-methyl-5-nitroimidazole-1-ethanol,anagiardil,trichazol,gineflavir,meronidal,metronidaz,novonidazol PubChem CID: 4173 ChEBI: CHEBI:6909 IUPAC Name: 2-(2-methyl-5-nitroimidazol-1-yl)ethanol SMILES: CC1=NC=C(N1CCO)[N+](=O)[O-]
Metronidazole, 99%
CAS: 443-48-1 Molecular Formula: C6H9N3O3 Molecular Weight (g/mol): 171.156 MDL Number: MFCD00009750 InChI Key: VAOCPAMSLUNLGC-UHFFFAOYSA-N Synonym: metronidazole,metronidazol,flagyl,2-methyl-5-nitroimidazole-1-ethanol,anagiardil,trichazol,gineflavir,meronidal,metronidaz,novonidazol PubChem CID: 4173 ChEBI: CHEBI:6909 IUPAC Name: 2-(2-methyl-5-nitroimidazol-1-yl)ethanol SMILES: CC1=NC=C(N1CCO)[N+](=O)[O-]
Metronidazole, 99.32%, MP Biomedicals™
CAS: 443-48-1 Molecular Formula: C6H9N3O3 Molecular Weight (g/mol): 171.156 InChI Key: VAOCPAMSLUNLGC-UHFFFAOYSA-N Synonym: metronidazole,metronidazol,flagyl,2-methyl-5-nitroimidazole-1-ethanol,anagiardil,trichazol,gineflavir,meronidal,metronidaz,novonidazol PubChem CID: 4173 ChEBI: CHEBI:6909 IUPAC Name: 2-(2-methyl-5-nitroimidazol-1-yl)ethanol SMILES: CC1=NC=C(N1CCO)[N+](=O)[O-]
Metronidazole, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Metronidazole, TRC
CAS: 443-48-1 Molecular Formula: C6 H9 N3 O3 Molecular Weight (g/mol): 171.15 Synonym: Metronidazole,Metronidazole Benzoate Imp. A (EP),2-Methyl-5-nitro-1H-imidazole-1-ethanol,1-(β-Hydroxyethyl)-2-methyl-5-nitroimidazole,1-(2-Hydroxyethyl)-2-methyl-5-nitroimidazole,1-Hydroxyethyl-2-methyl-5-nitroimidazole,2-Methyl-5-nitroimidazole-1-ethanol,Anagiardil,Arilin,Bayer 5360,Cimetrol 500LPCI,Clont,Deflamon,Efloran,Elyzol,Entizol,Flagesol,Flagil,Flagyl,Fossyol,Gineflavir,Klion,Klont,Medazol,Metro Cream & Gel,MetroGel,Metrogyl,Metrolag,Metrolyl,Metron,Metronidazol,Metropol,Metrotop,Mexibol,NSC 50364,NSC 69587,Nidazole,Noritate,Orvagil,Protostat,RP 8823,Rathimed,Sanatrichom,Satric,Takimetol,Trichazol,Tricho Cordes,Tricho-Gynaedron,Trichobrol,Trichocide,Trichomol,Trichopal,Tricocet,Trikhopol,Trivazol,Vagilen,Vagimid,Zadstat IUPAC Name: 2-(2-methyl-5-nitroimidazol-1-yl)ethanol SMILES: Cc1ncc([N+](=O)[O-])n1CCO
Metronidazole 2.0 mg/ml in Methanol, LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Metronidazole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Metronidazole, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality