accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2F

CAS RN 443-86-7

IUPAC Name: 3-fluoro-2-methylaniline
Synonyms: 3-fluoro-2-methyl-aniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: SLDLVGFPFFLYBM-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC=C1F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results