accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClOSOOHOCl

CAS RN 4430-20-0

IUPAC Name: 3,3-bis(3-chloro-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: chlorophenol red
Molecular Weight (g/mol): 423.26
Molecular Formula: C19H12Cl2O5S
InChi Key: WWAABJGNHFGXSJ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results