accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBrBrOSOOBrBrBrBrBrOHBr

CAS RN 4430-25-5

IUPAC Name: 4,5,6,7-tetrabromo-3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: tetrabromophenol blue
Molecular Weight (g/mol): 985.54
Molecular Formula: C19H6Br8O5S
InChi Key: QPMIVFWZGPTDPN-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC(Br)=C(O)C(Br)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results