CAS RN 4430-36-8
IUPAC Name:
1-isothiocyanato-4-(methylsulfanyl)butane
Synonyms:
1-isothiocyanato-4-(methylsulfanyl)butane
Molecular Weight (g/mol):
161.28
Molecular Formula:
C6H11NS2
InChi Key:
IHQDGXUYTSZGOG-UHFFFAOYSA-N
SMILES:
CSCCCCN=C=S
1
–
1
of
1
results
1
4-(Methylthiol)-1-(isothiocyanato)butane, TRC
CAS: 4430-36-8 Molecular Formula: C6 H11 N S2 Molecular Weight (g/mol): 161.29 Synonym: Erucin (4-(Methylthiol)-1-(Isothiocyanato)butane) IUPAC Name: 1-isothiocyanato-4-methylsulfanylbutane SMILES: CSCCCCN=C=S