accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4437-65-4

IUPAC Name: (4-methylquinolin-2-yl)sulfanide
Synonyms: (4-methylquinolin-2-yl)sulfanide
Molecular Weight (g/mol): 174.24
Molecular Formula: C10H8NS
InChi Key: OJOPTXKXKPMMGC-UHFFFAOYSA-M
SMILES: CC1=C2C=CC=CC2=NC([S-])=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results