accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FONNNNHSOONH2NH2F

CAS RN 443797-96-4

IUPAC Name: 4-{[5-amino-1-(2,6-difluorobenzoyl)-1H-1,2,4-triazol-3-yl]amino}benzene-1-sulfonamide
Synonyms: 4-{[5-amino-1-(2,6-difluorobenzoyl)-1,2,4-triazol-3-yl]amino}benzenesulfonamide
Molecular Weight (g/mol): 394.36
Molecular Formula: C15H12F2N6O3S
InChi Key: KDKUVYLMPJIGKA-UHFFFAOYSA-N
SMILES: NC1=NC(NC2=CC=C(C=C2)S(N)(=O)=O)=NN1C(=O)C1=C(F)C=CC=C1F
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results