accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHFFF

CAS RN 444-30-4

IUPAC Name: 2-(trifluoromethyl)phenol
Synonyms: O-trifluoromethylphenol
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: ZOQOPXVJANRGJZ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)