accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C

CAS RN 4445-07-2

IUPAC Name: octadecylbenzene
Synonyms: benzene, octadecyl-
Molecular Weight (g/mol): 330.6
Molecular Formula: C24H42
InChi Key: WSVDSBZMYJJMSB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results