accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH3NH3NH3NH3BrPdBr

CAS RN 44463-62-9

IUPAC Name: dibromopalladium tetraamine
Synonyms: palladium(II) bromide tetraamine
Molecular Weight (g/mol): 334.35
Molecular Formula: Br2H12N4Pd
InChi Key: BLPSMPASLSGGOC-UHFFFAOYSA-L
SMILES: N.N.N.N.Br[Pd]Br
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results