accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3HOCH3CH3HOCH3CH3HOGa

CAS RN 4452-61-3

IUPAC Name: tris(propan-2-ol) gallium
Synonyms: tris(isopropyl alcohol) gallium
Molecular Weight (g/mol): 250.01
Molecular Formula: C9H24GaO3
InChi Key: FUJNSPPLSODXIC-UHFFFAOYSA-N
SMILES: [Ga].CC(C)O.CC(C)O.CC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results