accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4455-77-0

H3COPO

CAS RN 4455-77-0

IUPAC Name: [(methoxymethyl)(phenyl)phosphoroso]benzene
Synonyms: [methoxymethyl(phenyl)phosphoroso]benzene
Molecular Weight (g/mol): 246.25
Molecular Formula: C14H15O2P
InChi Key: OEPKDBQOTLDTNC-UHFFFAOYSA-N
SMILES: COCP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)